Learn More
MOLECULAR DEPOT LLC Neuromedin U - 100 ug
Supplier:  MOLECULAR DEPOT LLC MDP0841
Encompass_Preferred
Neuromedin U (Human) is a synthetic peptide consisting of the sequence Phe-Arg-Val-Asp-Glu-Glu-Phe-Gln-Ser-Pro-Phe-Ala-Ser-Gln-Ser-Arg-Gly-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2 The molecular weight of this peptide is 3080 4 g/mol and the CAS number is 312306-89-1 This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13595