Learn More
MOLECULAR DEPOT LLC Neuromedin U Mouse - 500 ug
Supplier:  MOLECULAR DEPOT LLC MDP0842
Encompass_Preferred
Neuromedin U (Mouse) is a synthetic peptide consisting of the sequence Phe-Lys-Ala-Glu-Tyr-Gln-Ser-Pro-Ser-Val-Gly-Gln-Ser-Lys-Gly-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2 The molecular weight of this peptide is 2706 g/mol and the CAS number is 935658-91-6 This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13356