Learn More
Cusabio Technology LLC neuronal pentraxin I
Supplier:  Cusabio Technology LLC CSBE14336H
Encompass_Preferred
The Human Neuronal Pentraxin-1 (NPTX1) ELISA Kit is designed to quantitatively measure NPTX1 levels in human serum and plasma samples using a sandwich ELISA methodology This kit detects concentrations ranging from 7 8 pg/mL to 500 pg/mL with a sensitivity of 1 95 pg/mL ensuring precise quantification of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours offering flexibility for high-throughput applications Absorbance measurements at 450 nm enable reliable detection while the optimized protocol minimizes cross-reactivity and interference Ideal for research studies exploring neurological biomarkers this kit provides consistent reproducibility and complies with standard ELISA performance criteria for accurate analysis of NPTX1 in biological fluids
Catalog No. 50-000-48922