Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 1/EA
Supplier:  Biotium 29135
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Near-infrared CF 740 has Ex/Em at 742/767 nm 1 mg lyophilized solid
Catalog No. 50-005-01225