Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 5/PK
Supplier:  Biotium 29128
Encompass_Preferred
This product is fluorescently-labeled wheat germ agglutinin (WGA) WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine Near-infrared CF740 dye has Ex/Em at 742/767 nm 5x1mg
Catalog No. 50-005-01302