Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 1/EA
Supplier:  Biotium 29148
Encompass_Preferred
This product is fluorescently-labeled Ulex europaeus agglutinin I (UEA I) UEA I binds to glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates Near-infrared CF790 has Ex/Em at 783/808 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-01230