Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 5/PK
Supplier:  Biotium 29144
Encompass_Preferred
This product is fluorescently-labeled wheat germ agglutinin (WGA) WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine Near-infrared CF790 dye has Ex/Em at 783/808 nm 5x1mg
Catalog No. 50-005-00249