Learn More
CPC Scientific H-Glu-Ile-Gly-Asp-Glu-Glu-Asn-Ser-Ala-Lys-Phe-Pro-Ile-NH2 (ammonium salt) 5MG
SDP
Supplier:  CPC Scientific MCH002B
Encompass
NEI (neuropeptide EI) is processed from pro-MCH. It has been reported that rat MCH may act as a central corticotropin inhibitory factor involved in the circadian rhythmicity of plasma ACTH levels in the rat while NEI antagonizes its action.ONE-LETTER SEQUENCE: EIGEENSAKFPI-NH2MOLECULAR FORMULA: C63H98N16O23MOLECULAR WEIGHT:1447.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [125934-45-4], RESEARCH AREA: HormonalREFERENCES: M.-T.Bluet-Pajot et al., J. Neuroendocrinol., 7, 297 (1995)
Catalog No. 50-210-9722