Learn More
Medchemexpress LLC γ-Neuropeptide (rabbit) | 114882-65-4 | 99.99% | 2320.59 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP306910MG
Encompass_Preferred
γ-Neuropeptide (rabbit) is an endogenous neurokinin peptide isolated from rabbit intestine. It acts as a neurokinin 2 (NK2) receptor agonist and is involved in mediating the hypothalamic-pituitary-adrenal (HPA) axis and reproductive hormone release. This product is for research use only.
  • Acts as a neurokinin 2 (NK2) receptor agonist
  • Mediates the hypothalamic-pituitary-adrenal (HPA) axis
  • Involved in reproductive hormone release
  • Induces stimulation of LH release by hemipituitaries
  • For research use only
Catalog No. 50-003-98489