Learn More
Abcam Neuropeptide S (rat), NPS receptor agonist, 1MG
Supplier:  Abcam AB1202461MG
Encompass_Preferred
MW 2210.52 g/mol. Endogenous ligand for the NPS receptor involved in many biological functions including sleeping/wakening, locomotion, anxiety, and food intake.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7749