Learn More
Medchemexpress LLC Neuropilin-1 antibody (YA3507) | 100 UL
SDP
Supplier:  Medchemexpress LLC HYP83810100UL
Encompass_Preferred
The Neuropilin-1 antibody (YA3507) is a mouse-derived IgG1 monoclonal antibody targeting human Neuropilin-1. This antibody is affinity purified and has an observed molecular weight of 100 kDa. It plays a significant role in binding to VEGF-165, potentially inhibiting its binding to cells and inducing apoptosis. It may also interact with various members of the semaphorin family.
  • Mouse-derived IgG1 monoclonal antibody
  • Targets human Neuropilin-1
  • Affinity purified
  • Observed molecular weight of 100 kDa
  • Reacts with human samples
  • Suitable for WB, FC, and ELISA applications
  • Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles
Catalog No. 50-004-43349