Learn More
Abcam Neutral red, histology stain, 25G
Supplier:  Abcam AB14636525G
Encompass_Preferred
MW 288.78 g/mol. Commonly used histological stain for lysosymes that stains lysosomes red. Once taken up by cells the compound can be used to ascertain cell viability. Also used as a pH indicator.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4682