Learn More
Getinge USA Neutrawash Plu
Supplier:  Getinge USA 61301605890
Encompass
On-foaming liquid detergent for high performance cleaningSuperior cleaning abilityWorks well in the widest range of water conditionsFoam controlled and safe on all metals at use concentrations.
Catalog No. NC9628242
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.