Learn More
Cayman Chemical Neutrophil Myeloperoxidase Activity Assay Kit, 2 x 96 wells
SDP
Supplier:  Cayman Chemical 6006202X96WELLS
Encompass_Preferred
Myeloperoxidase (MPO), released from neutrophils to degrade invading pathogens, provides one of the earliest lines of defense in innate immunity. It catalyzes the hydrogen peroxide-mediated oxidation of halide ions to products such as hypochlorous acid (HOCl). These reactive products participate in a variety of secondary reactions which in turn lead to modifications of proteins and destruction of the extracellular matrix. Cayman’s Neutrophil Myeloperoxidase Activity Assay kit utilizes 3,3',5,5'-tetramethyl-benzidine (TMB) as a chromogenic substrate, which upon reacting with MPO, yields a blue color detectable by its absorbance at 650 nm. The color intensity is proportional to the amount of MPO in the sample. Reagents needed to isolate neutrophils from whole blood are included in the kit, as is PMA, which is known to stimulate MPO release from neutrophils. A specific inhibitor for MPO, 4-aminobenzhydrazide (ABH), is also included in the kit for verifying the specificity of the assay.
Catalog No. NC0810299