Learn More
Medchemexpress LLC Nevanimibe hydrochloride | 133825-81-7 | 99.5% | 458.08 g/mol | C27H40ClN3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100399A50MG
Encompass_Preferred
Nevanimibe hydrochloride is the hydrochloride salt of nevanimibe, an orally active and selective ACAT1 inhibitor supplied as a research-grade solid for in vitro and preclinical studies. The material is characterized by molecular identity and high purity suitable for biochemical and pharmacological assays.
  • High purity suitable for analytical and biological assays.
  • Hydrochloride salt form for improved stability and solubility in polar solvents.
  • Well-characterized molecular identity and molecular weight for reproducible results.
  • Available in small pack sizes appropriate for laboratory research.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-14568