Learn More
Medchemexpress LLC Nevirapine | 129618-40-2 | MFCD00866928 | 99.9% | 266.30 g/mol | C15H14N4O | 1 ML

Supplier: Medchemexpress LLC HY1057010MM 1ML
Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase supplied as a research reagent in a 10 mM solution (1 mL). It is intended for use in antiviral research, assay development, and analytical applications where a characterized NNRTI standard is required.
- Non-nucleoside reverse transcriptase inhibitor for HIV-1 research.
- Concentration: 10 mM in 1 mL solution.
- High purity: 99.9% assay.
- Chemical formula C15H14N4O; molecular weight 266.30 g/mol.
- Soluble in DMSO (≈53.66 mM) and compatible with common in vivo vehicles; refer to datasheet for preparation.
- Recommended storage conditions for powder and in-solution formats are provided on the product datasheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.