Learn More
Selleck Chemical LLC Nevirapine (NSC 641530) S1742-1g
SDP
Supplier:  Selleck Chemical LLC S17421G
Encompass_Preferred
Nevirapine (NSC 641530 NVP) is a non-nucleoside reverse transcriptase inhibitor (NNRTI) used to treat HIV-1 infection and AIDS
Catalog No. 50-311-0000