Learn More
Grainger New Dimension Stretch Dress Shirt L Navy
Supplier:  Grainger 14J254
Encompass_Preferred
The Grainger 14J254 is a 14-inch diameter, 254mm wide, Type 1A abrasive cutoff wheel made by Norton. It features aluminum oxide abrasive grains and is rated for use at 13,300 RPM with a maximum wheel speed of 19,500 surface feet per minute. This thin cutoff wheel is designed for straight cutting and side grinding of mild steel, stainless steel, aluminum and other metals. It should be used with a steel, aluminum oxide, or silicon carbide abrasive cutoff machine. This is a commercial-grade abrasive cutoff wheel made in the USA.
Catalog No. 50-275-2928