Learn More
Grainger New Dimension Stretch Dress Shirt L
Supplier:  Grainger 14K847
Encompass_Preferred
The Grainger 14K847 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 4mm to 14mm and 15 metric sockets ranging from 5/32 inch to 9/16 inch. The chrome vanadium sockets have a matte finish and feature a knurled design for easy hand tightening. They come packed inside a lightweight molded storage case for convenient portability and organization. This 30-piece socket set provides a versatile combination of standard and deep sockets for automotive repair and maintenance tasks.
Catalog No. 50-275-2705