Learn More
Grainger New Dimension Stretch Dress Shirt XL
Supplier:  Grainger 14K850
Encompass_Preferred
The Grainger 14K850 is a Westward brand 1/4-inch drive click torque wrench that measures torque in inch pounds. It has a range of 10 to 150 inch pounds and a +/- 4% accuracy. The wrench has a reversible ratchet head, is 15 inches long, and weighs 1.1 lbs. It has a knurled grip handle and a scale on the handle to set the desired torque value. The wrench clicks when the preset torque is reached. It can be calibrated and comes with a plastic storage case. This torque wrench is ideal for precisely tightening bolts and nuts in machinery, automotive, aviation and other applications.
Catalog No. 50-275-2706