Learn More
Braintree Scientific Mini Centrifuge, Red, Quantity: 1 each
SDP
Supplier:  Braintree Scientific C1601R
Encompass
A New Line of Mini Microcentrifuges The new mini micro-centrifuges will replace the current Spectrafuge™ Mini Centrifuges. Simple and convenient to operate, the new mini microcentrifuges feature greater capacity (8 place MCT and 4 place PCR strip rotors), a quick-release rotor system and electronic brake. Once samples area loaded and the lid is closed, the rotor rapidly accelerates to 6,000 rpm, making it an ideal centrifuge for bringing small droplets to the bottom of tubes for micro-filtrations or basic separations. Mini Microcentrifuge Product Features: Quick release rotor system - no tools needed to exchange rotors! Electronic braking - Allows for more rapid deceleration, less waiting Dual lid switch Compact, ruggeddesign Product Specifications Maximum Speed 6,000 rpm / 2,000 x g Maximum Capacity 8 x 1.5/2.0mL or 4 x 0.2mL strips Dimensions 6.8 x 5.8 x 4.8 in / 17.2 x 14.7 x 12.2 cm Electrical AC 100 - 240V, 50/60Hz
Catalog No. 50-195-4474