Learn More
IPHASE PHARMA SERVICES LLC New Zealand White Rabbit PBMC,Frozen,5million
Supplier:  IPHASE PHARMA SERVICES LLC 082I11.21
Encompass_Preferred
Peripheral Blood Mononuclear Cells (PBMCs) are a heterogeneous population of immune cells found in peripheral blood and isolated via density gradient centrifugation. They include lymphocytes—such as T cells, B cells, and natural killer (NK) cells—and monocytes.It can be applicated Immunological Research and Cell Therapy (CAR-T/CAR-NK). These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3967059