Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium hexacyanoferrate12KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 24402312KG
Encompass
Potassium hexacyanoferrate(III) (Potassium ferricyanide) can be prepared by the anodic oxidation of K4[Fe(CN)6].
Catalog No. 50-290-7494