Learn More
Alkali Scientific NextPette™ Pipettor, 1ml to 10ml
SDP
Supplier:  Alkali Scientific P970010M
Encompass_Preferred
NextPette's materials and components were selected to minimize while ensuring durability and long service life. The pipettors are designed with service in mind and are easy to disassemble, with seals that are simple to replace. Silicone lubricant is included with each pipettor. Recalibration can be done in the lab without tools, and the instruction manual provides guidance for checking pipetting accuracy and recalibrating when needed. NextPette™ variable volume pipette 0.1 to 1.0µL features an ergonomic design ideal for high volume laboratories. Adjust volume easily with one hand while viewing the large 4 digit display. Comfortable stroke distance suits all hand s. Individually tested for accuracy according to EN ISO 8655 and sold individually. Bulk Bag 250/pack for P9700 T5M and 100/pack for P9700 T10M. Macro Set P9700 MARCO includes P9700 20 (20–200ul), P9700 1000 (100–1000ul), and P9700 5M (1–5ml).
Catalog No. 50-006-62101