Learn More
TARGETMOL CHEMICALS INC NF-56-EJ40 HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T12216L25MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. NF-56-EJ40 hydrochloride is a potent high-affinity and highly selective human SUCNR1 (GPR91) antagonist with an IC50 of 25 nM and a Ki of 33 nM and shows almost no activity towards rat SUCNR1. NF-56-EJ40 hydrochloride has high affinity for humanized rat SUCNR1 with a Ki value of 17.4 nM. purity: 97%
Catalog No. 50-257-7430