Learn More
Abcam NF kappaB p65 (pS536) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1766471X96TESTS
Encompass_Preferred
NF kappaB p65 (pS536) ELISA Kit is a single-wash 90-min Simplestep used to quantify NF kappaB p65 (pS536) with a sensitivity of 5 µg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 15 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2295