Learn More
System Biosciences LLC NF-KB/JURKAT/GFP CELL LINE
SDP
Supplier:  System Biosciences LLC TR850A1
Encompass_Preferred
NF-κB/Jurkat/GFP™ Transcriptional Reporter Cell Line. Speed your studies with this pre-built Jurkat cell line for monitoring NF-κB signaling in real time. We’ve already integrated an expression cassette that includes NF-κB-responsive transcriptional elements upstream of a minimal CMV promoter (mCMV)-GFP cassette. Expression of GFP (up to 30-fold over background) only occurs in the presence of active NF-κB signaling, enabling screening for genetic and/or small molecule inhibitors and activators of the NF-κB signaling pathway.
Catalog No. NC0687295