Learn More
Medchemexpress LLC NF-KB p105/p50 Antibody (YA699) | Protein L affinity purified | Predicted band size: 105 kDa; Observed band size: 50/120 kDa | 50 UL

Supplier: Medchemexpress LLC HYP8024450UL
The NF-KB p105/p50 Antibody (YA699) is a Mouse-derived, non-conjugated IgM monoclonal antibody designed for research use. It targets NF-KB p105/p50, a pleiotropic transcription factor involved in various biological processes such as inflammation, immunity, differentiation, cell growth, tumorigenesis, and apoptosis.
- Host: Mouse
- Clonality: Monoclonal
- Molecular weight: Predicted 105 kDa; Observed 50/120 kDa
- Species reactivity: Human, mouse
- Purity: Protein L affinity purified
- Conjugation: Non-conjugated
- Isotype: IgM
- Formulation: Supplied in 1*PBS (pH7.4), 0.2% BSA, 40% Glycerol, 0.05% Sodium Azide
- Appearance: Liquid
- Applications: WB (1:1000-1:2000), ICC/IF (1:200), IHC-P (1:200-1:500)
- Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.