Learn More
Medchemexpress LLC NF-KB p105/p50 Antibody (YA700) | 100 UL
SDP
Supplier:  Medchemexpress LLC HYP80764100UL
Encompass_Preferred
NF-KB p105/p50 Antibody (YA700) is a mouse-derived, non-conjugated IgG1 monoclonal antibody that targets NF-KB p105/p50. It is intended for research use only and plays a crucial role in studying various biological processes such as inflammation, immunity, differentiation, cell growth, tumorigenesis, and apoptosis.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets NF-KB p105/p50
  • Intended for research use only
  • Involved in inflammation, immunity, differentiation, cell growth, tumorigenesis, and apoptosis
  • Facilitates cytoplasmic retention of NF-kappa-B proteins by p105 and generation of p50 subunit
  • P50 subunit binds to kappa-B consensus sequence in enhancer regions of genes involved in immune response
  • Precursor of the active p50 subunit
Catalog No. 50-004-42272