Learn More
Cayman Chemical NF449sodIum salt 5mg
SDP
Supplier:  Cayman Chemical 133245 MG
Encompass_Preferred
An analog of suramin that selectively inhibits P2X1 receptors (pIC50 = 6.3); selectively antagonizes the Gsα-subunit G protein, which suppresses the association rate of GTPγS binding to Gsα-s, inhibits the stimulation of adenylyl cyclase activity, and blocks G protein coupling to certain GPCRs
Catalog No. 50-295-2551