Learn More
ATCC NF639; Mammary Tumor; Mouse (Mus musculus)
Supplier:  ATCC CRL3090
Encompass
NF639 is an epithelial cell line isolated from the mammary glands of a female mouse carrying the MMTV-c-neu transgene. The cell line NF639 overexpresses the oncogene: neu. It is useful in the studies of the activation and requirement for EGFR2 signal transduction pathways in mammary gland tumors.
Catalog No. 50-238-3175