Learn More
Santa Cruz Biotechnology NFAT ANTIBODY
Supplier:  Santa Cruz Biotechnology SC7294
Encompass
NFATc1 (7A6), mouse monoclonal IgG1; 200microgram/ml, epitope corresponding to amino acids 197-304 of NFATc1 of human origin, specific for NFATc1 (also designated NFATc), mouse and human reactive, Western blotting
Catalog No. NC9890017