Learn More
Abeomics Nfat Reporter Hek293 Cell Line
Supplier:  Abeomics 14110ACL
Encompass_Preferred
The NFAT Leeporter Luciferase Reporter cell line is a stably transfected HEK 293 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the Nuclear Factor of Activated T-cells (NFAT) response element, so that the cell line is designed to measure the transcriptional activity of NFAT. NFAT is a transcription factor originally found in activated T lymphocytes, and is now known to regulate not only T cell activation and differentiation but also the function of other immune cells including dendritic cells, B cells and megakaryocytes. Cell line application: (1) Monitor the NFAT signaling pathway activity, and (2) Screen for activators or inhibitors of the NFAT signaling pathway. .
Catalog No. NC1745315