Learn More
TARGETMOL CHEMICALS INC NFATC1-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T615315MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. NFATc1-IN-1 (also known as compound A04) is a highly effective inhibitor of osteoclast formation induced by RANKL with an IC50 of 1.57 uM. It exerts its anti-osteoclastogenic effects by attenuating the RANKL-induced nuclear translocation of NFATc1. Due to its remarkable properties NFATc1-IN-1 holds significant potential for advancing research related to osteoclastic diseases [1]. purity: 99%
Catalog No. 50-257-9514