Learn More
ABclonal Technology NFE2L3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A157611000UL
Encompass_Preferred
This gene encodes a member of the cap n collar basic-region leucine zipper family of transcription factors. The encoded protein heterodimerizes with small musculoaponeurotic fibrosarcoma factors to bind antioxidant response elements in target genes. This protein is a membrane bound glycoprotein that is targeted to the endoplasmic reticulum and the nuclear envelope. Pseudogenes of this gene are found on chromosomes 16, 17, and 18.
Catalog No. 50-001-05467