Learn More
Cayman Chemical NFKB ActIvatIon InHBTr II 25mg
SDP
Supplier:  Cayman Chemical 1908325 MG
Encompass_Preferred
An inhibitor of NF-κB activation; inhibits TNF-α-induced NF-κB activation in HT-1080 cells at 20 µM; inhibits TNF-α-induced expression of MMP-9 in HT-1080 cells; decreases TNF-α-induced migration of HT-1080 cells at 20 µM; inhibits IL-6 release induced by TNF-α from primary rat brain pericytes at 1, 5, and 10 µM; increases IL-8 secretion in HT-29 and Caco-2 cells at 5 µM in combination with glyoxal
Catalog No. 50-298-3600