Learn More
Gentarget NFKB-LUC /GFP/ LENTIVIRUS IN P
Supplier:  Gentarget LVP967GPBS
Encompass
Pre-made lentivirus express Firefly Luciferase 3 reporter under a minimal promoter inserted with 4 tandem repeats of NFκB transcriptional response element /NFkB-TRE/ for detecting NFkB regulated signaling pathway activity.
Catalog No. NC3711043