Learn More
Aviva Systems Biology Corp NFKBID Antibody - C-terminal region (ARP60648_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60648_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKBID. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NFKBID. Alternate names: IkappaBNS, MGC11314, MGC149503, TA-NFKBHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-115