Learn More
Cayman Chemical NG NG-dImethyl-L-ArgInIn 5mg
SDP
Supplier:  Cayman Chemical 335765 MG
Encompass_Preferred
An endogenous NOS inhibitor; levels are increased in Dahl salt-sensitive rats fed a high-salt diet concomittant with an increase in blood pressure
Catalog No. 50-294-6504