Learn More
AdipoGen NG 012 (1 mg)
SDP
Supplier:  AdipoGen AGCN20155M001
Encompass_Preferred
Chemical. CAS: 141731-76-2. Formula: C32H38O15. MW: 662.7. Isolated from Penicillium sp. FKI-5387. Antibiotic. Antifungal. Nerve growth factor (NGF) potentiator.
Catalog No. 50-205-3538