Learn More
TARGETMOL CHEMICALS INC NG25 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T56435MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NG25 is a potent dual TAK1 and MAP4K2 inhibitor with IC50s of 149 nM and 21.7 nM respectively. purity: 99%
Catalog No. 50-257-9742