Learn More
Cayman Chemical NGaminoLArginine hydrochlori
SDP
Supplier:  Cayman Chemical 105545 MG
Encompass_Preferred
Inhibits nNOS, iNOS, and eNOS (Ki = 0.3, 3, and 2.5 μM, respectively); can be used both in cell culture and in vivo
Catalog No. 50-288-8978