Learn More
Medchemexpress LLC NGB 2904 | 189060-98-8 | 98.3% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1269710MG
Encompass_Preferred
NGB 2904 is a potent, selective, orally active, and brain-penetrated antagonist of the dopamine D3 receptor. It exhibits high selectivity for the D3 receptor over D2, 5-HT2, α1, D4, D1, and D5 receptors. This compound also antagonizes Quinpirole-stimulated mitogenesis.
  • Potent and selective dopamine D3 receptor antagonist
  • Orally active and brain-penetrated
  • High selectivity over other dopamine and serotonin receptors
  • Antagonizes Quinpirole-stimulated mitogenesis
Catalog No. 50-003-38415