Learn More
Cayman Chemical ngenol-3-ngelate 10mg
SDP
Supplier:  Cayman Chemical 1620710 MG
Encompass_Preferred
A hydrophobic diterpene ester that rapidly induces cell death in proliferating keratinocytes through plasma membrane and mitochondrial disruption; causes inflammation due, at least in part, to activation of PKC, leading to antibody-dependent cellular cytotoxicity
Catalog No. 50-296-9625