Learn More
Cayman Chemical ngIotensIn II InhIbItr50mL
SDP
Supplier:  Cayman Chemical 900068150 ML
Encompass_Preferred
An angiotensin II (AII) inhibition cocktail has been developed by Bertin Pharma in order to fix physiological concentration of AII in biological samples
Catalog No. 50-289-3976