Learn More
Cayman Chemical ngIotensIn IIIhumnmouse 10mg
SDP
Supplier:  Cayman Chemical 2161810 MG
Encompass_Preferred
An agonist at AT1 and AT2 receptors, with a 30-fold higher affinity for AT2
Catalog No. 50-296-4237