Learn More
Cayman Chemical ngIotensIn IIIhumnmouse 5mg
SDP
Supplier:  Cayman Chemical 216185 MG
Encompass_Preferred
An agonist at AT1 and AT2 receptors, with a 30-fold higher affinity for AT2
Catalog No. 50-298-6319