Learn More
ABclonal Technology NGLY1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2276120UL
Encompass_Preferred
This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-05886