Learn More
Meridian Life Science NGS Clean and Select Beads, 500ML
SDP
Supplier:  Meridian Life Science MDX041500
Encompass_Preferred
NGS Clean and Select Beads is an efficient NGS library preparation clean-up and selection system based on paramagnetic SPRI beads. It is designed to efficiently removing contaminants, such as nucleotides, primers, adapters, enzymes, buffer additives and salts, and for size selection of DNA fragments that can be used directly for downstream applications. The protocol can be used manually or adapted for liquid handling workstations.
Catalog No. 50-008-56494