Learn More
Abcam NH 125, CaMK III inhibitor, 50MG
Supplier:  Abcam AB14518850MG
Encompass_Preferred
MW 524.6 Da, Purity >99%. Potent, selective CaMK III inhibitor (IC₅₀ = 60 nM). Histidine protein kinase inhibitor (IC₅₀ = 6.6 μM). Induces G1/S cell cycle arrest and apoptosis. Shows anticancer effects. Antibacterial agent in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5418